missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PDE6C Recombinant Mouse Monoclonal Antibody (K92073_15A1)

Product Code. 30830398
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30830398 50 μL
30824806 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30830398 Supplier Invitrogen™ Supplier No. MA565945

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Recombinant Monoclonal Antibody

Immunogen sequence: QILMWSANKV FEELTDVERQ FHKALYTVRS YLNCERYSIG LLDMTKEKEF YDEWPIKLGE VEPYKGPKTP DGREVNFYKI IDYILHGKEE IKVIPTPPAD HWTLISGLPT YVAENGFICN MMNAPADEYF TFQKGPVDET GWVIKNVLSL PIVNKKEDIV GVATFYNRKD GKPFDEHDEY ITETLTQFLG WSLLNTDTYD KMNKLENRKD IAQEMLMNQT Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 29%, Pig - 95%, Cynomolgus monkey - 99%.

PDE6C is the alpha-prime subunit of cone phosphodiesterase, which is composed of a homodimer of two alpha-prime subunits and 3 smaller proteins of 11, 13, and 15 kDa.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PDE6C
Applications Immunohistochemistry (Paraffin)
Classification Recombinant Monoclonal
Clone K92073_15A1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.5% BSA, 50% glycerol and 0.09% sodium azide
Gene PDE6C
Gene Accession No. P51160
Gene Alias ACHM5; cGMP phosphodiesterase 6C; COD4; Cone cGMP-specific 3',5'-cyclic phosphodiesterase subunit alpha'; cpfl1; PDE6C; PDEA2; phosphodiesterase 6C; phosphodiesterase 6C, cGMP specific, cone, alpha prime; phosphodiesterase 6C, cGMP-specific, cone, alpha prime
Gene Symbols PDE6C
Host Species Mouse
Immunogen Recombinant human PDE6C (aa 241-460).
Purification Method Protein A
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5146
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.