missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDGF-C Rabbit anti-Human, Clone: 4, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89373-100UL
This item is not returnable.
View return policy
Description
PDGF-C Monoclonal specifically detects PDGF-C in Human samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| PDGF-C | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA, Immunofluorescence | |
| 4 | |
| ELISA 1:5000-1:10000, Immunocytochemistry/Immunofluorescence 1:20-1:100 | |
| FALLOTEIN, PDGF-C, platelet derived growth factor C, platelet-derived growth factor C, SCDGFSpinal cord-derived growth factor, secretory growth factor-like protein, VEGF-E | |
| Recombinant Human PDGF-C Protein | |
| 100 μg | |
| Cell Cycle and Replication | |
| 56034 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction