missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDGFA Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
227.00 € - 326.00 €
Specifications
| Antigen | PDGFA |
|---|---|
| Dilution | Western Blot 1:500-1:2000, Immunoprecipitation 0.5-2 uL/mg of lysate |
| Applications | Western Blot, Immunoprecipitation |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18674665
|
Novus Biologicals
NBP2-99113-100ul |
100 μg |
326.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18625045
|
Novus Biologicals
NBP2-99113-50ul |
50 μg |
227.00 €
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PDGFA Polyclonal antibody specifically detects PDGFA in Human samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
| PDGFA | |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Cell Cycle and Replication | |
| 0.2 um filtered solution in PBS | |
| 5154 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Western Blot 1:500-1:2000, Immunoprecipitation 0.5-2 uL/mg of lysate | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| PDGF-1, PDGF1PDGF A-chain, PDGF-A, Platelet-derived growth factor A chain, platelet-derived growth factor alpha chain, platelet-derived growth factor alpha isoform 2 preproprotein, platelet-derived growth factor alpha polypeptidePDGF subunit A, platelet-derived growth factor subunit A | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human PDGFA. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title