missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDGFRB(phospho Y740) & PDGFRB Protein Phosphorylation Antibody Pair, Abnova™
Description
This protein phosphorylation antibody pair set comes with two antibodies, one against the PDGFRB protein, and the other against the specific Y740 phosphorylated site of PDGFRB for use in in situ Proximity Ligation Assay.
Specifications
Specifications
| Antigen | PDGFRB |
| Applications | In situ PLA |
| Conjugate | Unconjugated |
| Gene | platelet-derived growth factor receptor, beta polypeptide |
| Gene Alias | CD140B/JTK12/PDGF-R-beta/PDGFR/PDGFR1 |
| Gene Symbols | PDGFRB |
| Quantity | 1 Set |
| Regulatory Status | RUO |
| Primary or Secondary | Primary |
| Gene ID (Entrez) | 5159 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?