missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDX-1/IPF1 Antibody (267712), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB2419P
This item is not returnable.
View return policy
Description
PDX-1/IPF1 Monoclonal antibody specifically detects PDX-1/IPF1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Flow Cytometry, Immunocytochemistry
Specifications
| PDX-1/IPF1 | |
| Monoclonal | |
| PE | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, Immunohistochemistry, Flow Cytometry, Immunocytochemistry | |
| 267712 | |
| Western Blot, Immunohistochemistry, Intracellular Staining by Flow Cytometry, Immunocytochemistry | |
| Glucose-sensitive factor, IDX-1GSF, Insulin promoter factor 1, insulin promoter factor 1, homeodomain transcription factor, Insulin upstream factor 1, IPF1pancreas/duodenum homeobox protein 1, Islet/duodenum homeobox-1, MODY4IUF1, pancreatic and duodenal homeobox 1, pancreatic-duodenal homeobox factor 1, PDX-1IPF-1, somatostatin transcription factor 1, Somatostatin-transactivating factor 1, STF-1IUF-1 | |
| E. coli-derived recombinant human PDX-1, Ala91-Arg283, Accession # P52945 | |
| 0.1 mL | |
| Lipid and Metabolism, Stem Cell Markers | |
| 3651 | |
| Store at 4C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction