missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PDX1 Recombinant Rabbit Monoclonal Antibody (5H2B1)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557981
This item is not returnable.
View return policy
Description
Immunogen sequence: NLTERHIKIW FQNRRMKWKK EEDKKRGGGT AVGGGGVAEP EQDCAVTSGE ELLALPPPPP PGGAVPPAAP VAAREGRLPP GLSASPQPSS VAPRRPQEPR.
The protein encoded by this gene is a transcriptional activator of several genes, including insulin, somatostatin, glucokinase, islet amyloid polypeptide, and glucose transporter type 2. The encoded nuclear protein is involved in the early development of the pancreas and plays a major role in glucose-dependent regulation of insulin gene expression. Defects in this gene are a cause of pancreatic agenesis, which can lead to early-onset insulin-dependent diabetes mellitus (NIDDM), as well as maturity onset diabetes of the young type 4 (MODY4).
Specifications
| PDX1 | |
| Recombinant Monoclonal | |
| 1.32 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| P52946, P52947 | |
| PDX1 | |
| A synthetic peptide corresponding to a sequence within amino acids 184-283 of human PDX1 (NP_000200.1). | |
| 20 μL | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| 5H2B1 | |
| Unconjugated | |
| PDX1 | |
| DLDBP; E3BP; Glucose-sensitive factor; GSF; Idx1; IDX-1; Insulin promoter factor 1; insulin promoter factor 1, homeodomain transcription factor; insulin promotor factor 1; insulin upstream factor 1; Ipf1; IPF-1; Islet/duodenum homeobox 1; islet/duodenum homeobox-1; IUF1; IUF-1; MODY4; PAGEN1; Pancreas/duodenum homeobox protein 1; pancreatic and duodenal homeobox 1; pancreatic and duodenal homeobox gene 1; pancreatic-duodenal homeobox factor 1; PDHX; PDX1; pdx-1; somatostatin transactivating factor 1; somatostatin transcription factor 1; somatostatin-transactivating factor 1; Stf1; STF-1 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 18609, 29535 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction