missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDZK1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 4 publications
237.00 € - 473.00 €
Specifications
| Antigen | PDZK1 |
|---|---|
| Dilution | Western Blot 1:1000, Simple Western 1:250, Immunohistochemistry 1:400, Immunocytochemistry/Immunofluorescence 1:200, Immunohistochemistry-Paraffin 1:400 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18062894
|
Novus Biologicals
NB400-149SS |
0.025 mL |
237.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18788603
|
Novus Biologicals
NB400-149 |
473.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
PDZK1 Polyclonal specifically detects PDZK1 in Human, Mouse, Bovine, Rat (Negative) samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| PDZK1 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cellular Markers, Diabetes Research, Endothelial Cell Markers, Growth and Development, Lipid and Metabolism, Neuronal Cell Markers, Plasma Membrane Markers, Signal Transduction | |
| Whole antisera with 0.05% Sodium Azide | |
| CAP70, CFTR-associated protein of 70 kDa, CLAMP, Na(+)/H(+) exchanger regulatory factor 3, Na/Pi cotransporter C-terminal-associated protein 1, naPi-Cap1, NHERF-3, NHERF3PDZD1Na(+)/H(+) exchange regulatory cofactor NHE-RF3, PDZ domain containing 1, PDZ domain-containing protein 1, PDZ-containing kidney protein 1, Sodium-hydrogen exchanger regulatory factor 3 | |
| PDZK1 | |
| IgG | |
| Unpurified | |
| Human, Bovine and mouse. Does not react with rat. |
| Western Blot 1:1000, Simple Western 1:250, Immunohistochemistry 1:400, Immunocytochemistry/Immunofluorescence 1:200, Immunohistochemistry-Paraffin 1:400 | |
| Polyclonal | |
| Antisera | |
| RUO | |
| Human, Mouse, Bovine, Rat (Negative) | |
| Q5T2W1 | |
| 5174 | |
| A synthetic peptide made to the C-terminus of human PDZK1. [UniProt# Q13113] | |
| Primary | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| 57 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title