missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PEA-15 Antibody, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-05543-100ul
This item is not returnable.
View return policy
Description
PEA-15 Monoclonal antibody specifically detects PEA-15 in Human, Mouse, Rat, Bovine samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| PEA-15 | |
| Monoclonal | |
| Unconjugated | |
| 50% PBS, 50% glycerol | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Bovine | |
| Purified |
| Western Blot, Immunofluorescence | |
| 1 mg/mL | |
| Western Blot 1:1000-1:2000, Immunocytochemistry/ Immunofluorescence 1:1000-2000 | |
| HMAT115kD, PEA-15, PED15 kDa phosphoprotein enriched in astrocytes, phosphoprotein enriched in astrocytes 15, Phosphoprotein enriched in diabetes | |
| Full length human PEA-15 as expressed in and purified from E. coli. | |
| 100 μg | |
| Apoptosis, Cell Biology, Cellular Markers | |
| 8682 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction