missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PEBP1 Recombinant Rabbit Monoclonal Antibody (24GB860)

Rabbit Recombinant Monoclonal Antibody
198.00 € - 564.00 €
Specifications
| Antigen | PEBP1 |
|---|---|
| Clone | 24GB860 |
| Content And Storage | -20°C |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30274851
![]() |
Invitrogen™
MA557538 |
100 μL |
564.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30276328
|
Invitrogen™
MA557539 |
20 μL |
198.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PEBP1 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
PBP binds ATP, opioids and phosphatidylethanolamine, exhibiting a lower affinity for phosphatidylinositol and phosphatidylcholine. This serine protease inhibitor inhibits thrombin, neuropsin and chymotrypsin but not trypsin, tissue type plasminogen activator and elastase. PBP contains hippocampal cholinergic neurostimulating peptide (HCNP), which may be involved in the function of the presynaptic cholinergic neurons of the central nervous system. HCNP increases the production of choline acetyltransferase but not acetylcholinesterase.Specifications
| PEBP1 | |
| -20°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human, Mouse, Rat | |
| PEBP1 | |
| 23 kDa morphine-binding protein; Basic cytosolic 21 kDa protein; epididymis luminal protein 210; epididymis secretory protein Li 34; epididymis secretory protein Li 96; HCNP; HCNPpp; HEL-210; HEL-S-34; HEL-S-96; Hippocampal cholinergic neurostimulating peptide; KIAA0299; LDGF; MDGF; MOCA; neuropolypeptide h3; P23K; PBP; Pbp1; Pbpr; PEBP; PEBP1; PEBP-1; phosphatidylethanolamine binding protein; phosphatidylethanolamine binding protein 1; phosphatidylethanolamine-binding protein; phosphatidylethanolamine-binding protein 1; PPBP; prostatic binding protein; Prostatic-binding protein; Raf kinase inhibitor protein; Raf kinase inhibitory protein; Raf-1 inhibitor protein; Raf-1 kinase inhibitor protein; RKIP; similar to phosphatidylethanolamine binding protein; zgc:56033; zgc:76942 | |
| PEBP1 | |
| Primary | |
| Affinity Chromatography |
| 24GB860 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P30086, P31044, P70296 | |
| 23980, 29542, 5037 | |
| A synthesized peptide derived from human PEBP1 (aa 64-99). | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title