missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PECR Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93014-0.02ml
This item is not returnable.
View return policy
Description
PECR Polyclonal antibody specifically detects PECR in Mouse samples. It is validated for Western Blot
Specifications
| PECR | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| DCR-RP, EC 1.3.1.38, HPDHASE, HSA250303, peroxisomal trans-2-enoyl-CoA reductase, putative short chain alcohol dehydrogenase, pVI-ARL, PVIARL2,4-dienoyl-CoA reductase-related protein, SDR29C1, short chain dehydrogenase/reductase family 29C, member 1, TERPDCRRP | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 24-303 of human PECR (NP_060911.2). TGGATGIGKAIVKELLELGSNVVIASRKLERLKSAADELQANLPPTKQARVIPIQCNIRNEEEVNNLVKSTLDTFGKINFLVNNGGGQFLSPAEHISSKGWHAVLETNLTGTFYMCKAVYSSWMKEHGGSIVNIIVPTKAGFPLAVHSGAARAGVYNLTKSLALEWACSGIRINCVAPGVIYSQTAVENYGSWGQSFFEGSFQKIPAKRIGVPEEVSSVVCFLLSPAASFITGQSVDVDGGRSLYTHSYEVPDHDNWPKGAGDLSVVKKMKETFKEKAKL | |
| 0.02 mL | |
| Apoptosis | |
| 55825 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction