missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Total Carbonyl Assay Kit (Colorimetric)

Product Code. 18605000 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 Assays
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18605000 100 Assays 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18605000 Supplier R&D Systems™ Supplier No. NBP325829
  • ONLINE EXCLUSIVE PRICE
    393.90 €
    329.70 € / 1 sets
    Save 64.20 €
    16% Off
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Assay Kit (Colorimetric)

  • Designed for spectrophotometric detection.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Storage of components varies. See protocol for specific instructions.
Detection Method Colorimetric
Product Line Novus
Quantity 100 Assays
Type Assay
Species Non-species specific
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.