Learn More
PEG3 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 23569-1-AP-20UL
 View more versions of this product
Product Code. 18014735
  • 145.00 € / 20µL
Description
The PEG3 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human PEG3. This antibody recognizes human, mouse antigen. The PEG3 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
PEG3
900 μg/mL
Rabbit
Primary
Human, Mouse
150 kDa
IgG
Western Blot, Immunohistochemistry, ELISA
KIAA0287, paternally expressed 3, PEG3, PW1, ZSCAN24
20 μL
5178
Store at -20°C. Stable for one year after shipment.