missing translation for 'onlineSavingsMsg'
Learn More

Melford Penicillin G, Sodium Salt

Product Code. 30018741
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30018741 5 g 5g
30018740 25 g 25g
30018739 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30018741 Supplier Melford Supplier No. P930005.0
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Penicillin G sodium is the sodium salt form of penicillin G, which is a narrow-spectrum beta-lactam antibiotic.

Penicillin G sodium is the sodium salt form of penicillin G, which is a narrow-spectrum beta-lactam antibiotic. It is one of the earliest and most widely used antibiotics, belonging to the penicillin class of antibiotics.

Penicillin G sodium interferes with bacterial cell wall synthesis. It inhibits the enzyme transpeptidase, which is involved in the cross-linking of peptidoglycan chains in the bacterial cell wall. This disruption weakens the cell wall structure, leading to cell lysis and death.

Also, Penicillin G is effective against a wide range of Gram-positive bacteria, including Streptococcus, Staphylococcus, and some anaerobic bacteria. It is less effective against Gram-negative bacteria.

Often used as a selective agent in culture media to isolate bacteria that are sensitive to this antibiotic. Its inclusion in culture medium allows for the specific growth of bacteria lacking resistance to penicillin G.

Specifications

Chemical Name or Material Penicillin G, Sodium Salt
CAS 69-57-8
Quantity 5 g
Molecular Formula C16H17N2NaO4S
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.