missing translation for 'onlineSavingsMsg'
Learn More

Penicillin G Sodium Salt, Fisher BioReagents

Product Code. 10790340
missing translation for 'orderingAttributeHoverText'
Quantity:
5 g
missing translation for 'unitSize'
5g
This item is not returnable. View return policy

Product Code. 10790340

missing translation for 'mfr': Fisher BioReagents BP29555

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Interferes with peptidoglycan synthesis in bacterial cell walls

TRUSTED_SUSTAINABILITY

Specifications

Type Penicillin G Sodium Salt
Physical Form Powder/Solid
CAS 69-57-8
pH 5.0 to 6.5 (at 30g/L aq.sol.)
Source Penicillium fungi
Loss on Drying < 1.5%
Packaging Amber Glass
Potency 1500 to 1750 U/mg
Quantity 5 g
Molecular Formula C16H17N2NaO4S
Merck Index 15, 7203
InChI Key FCPVYOBCFFNJFS-LQDWTQKMSA-M
SMILES CC1(C(N2C(S1)C(C2=O)NC(=O)CC3=CC=CC=C3)C(=O)[O-])C.[Na+]
IUPAC Name sodium;(2S,5R,6R)-3,3-dimethyl-7-oxo-6-[(2-phenylacetyl)amino]-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate
Molecular Weight (g/mol) 356.372
PubChem CID 23668834
ChEBI CHEBI:51765
Formula Weight 356.4
Chemical Name or Material Penicillin G Sodium Salt
Show More Show Less
compliance-icons
Product Identifier
  • Penicillin G sodium salt
Signal Word
  • Danger
Hazard Category
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.