missing translation for 'onlineSavingsMsg'
Learn More

Penicillin G sodium salt, MedChemExpress

Product Code. 16478766
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16478766 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16478766 Supplier Medchem Express Supplier No. HYB1463
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Penicillin G sodium salt is a typical β-lactam antibiotic.

Penicillin G sodium salt is a typical β-lactam antibiotic.

Specifications

Chemical Name or Material Penicillin G sodium salt
Physical Form Solid
CAS 69-57-8
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C16H17N2NaO4S
For Use With (Application) COVID-19-immunoregulation
Synonym Benzylpenicillin sodium salt
Solubility Information DMSO : ≥ 100 mg/mL (280.61 mM) ∣H2O : ≥ 100 mg/mL (280.61 mM)
SMILES O=C([C@@H](C(C)(C)S[C@]1([H])[C@@H]2NC(CC3=CC=CC=C3)=O)N1C2=O)O[Na]
Molecular Weight (g/mol) 356.37
Formula Weight 356.37
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.