missing translation for 'onlineSavingsMsg'
Learn More

Penicillin G , Sodium Salt, MP Biomedicals

Product Code. 17610937 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 mU
Unit Size:
10 micro units
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17610937 10 mU 10 micro units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17610937 Supplier MP Biomedicals Supplier No. 0219453710
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

An antibiotic that inhibits bacterial cell wall synthesis by binding to penicillin-binding proteins.

  • Penicillin G is a narrow spectrum antibiotic derived from Streptococcus pneumonia
  • It has been used to study the diagnostic and therapeutic implications of gentamicin-resistant Enterococcus faecalis sequence type 6 with reduced penicillin susceptibility
  • Also used in cell culture both alone and combined with streptomycin and other antibiotics

Specifications

Type Penicillin G Sodium Salt
Physical Form Powder
CAS 69-57-8
Color White
Quantity 10 mU
Molecular Formula C16H17N2NaO4S
Linear Formula C16H17N2NaSO4
MDL Number MFCD00069666
Synonym penicillin g sodium, penicillin g sodium salt, crystapen, benzylpenicillin sodium, american penicillin, sodium penicillin g, benzylpenicillin sodium salt, sodium penicillin, pencillin g sodium, sodium benzylpenicillinate
Solubility Information Soluble in water(H
InChI Key FCPVYOBCFFNJFS-LQDWTQKMSA-M
SMILES CC1(C(N2C(S1)C(C2=O)NC(=O)CC3=CC=CC=C3)C(=O)[O-])C.[Na+]
IUPAC Name sodium;(2S,5R,6R)-3,3-dimethyl-7-oxo-6-[(2-phenylacetyl)amino]-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate
Molecular Weight (g/mol) 356.372
PubChem CID 23668834
ChEBI CHEBI:51765
Formula Weight 356.38
Grade Cell Culture
Chemical Name or Material PENICILLIN G , SODIUM SALT
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Penicillin G Sodium Salt, cell culture grade
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-[In case of inadequate ventilation] wear respiratory protection.
  • P321-Specific treatment (see on this label).
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Penicillin G sodiumsalt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.