Learn More
Penicillin G sodium salt
Description
Penicillin G sodium salt is used as a β-lactam antibiotic. Penicillin G can be used as a selective agent in several types of isolation media. It has been used to study the diagnostic and therapeutic implications of gentamicin-resistant Enterococcus faecalis sequence type 6 with reduced penicillin susceptibility and in cell culture both alone and combined with streptomycin and other antibiotics.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
- Penicillin G sodium salt is the sodium salt of benzylpenicillin, a broad-spectrum β-lactam antibiotic with gram-positive and gram-negative bactericidal activity
- Penicillin G binds and inactivates penicillin-binding-proteins associated with the bacterial cell wall, thus inhibiting cell wall synthesis and resulting in cell lysis
Applications
- Penicillin G can be used as a selection agent in the isolation of microorganisms or in cell culture applications
- It can also be used to prevent infection in research animals
Specifications
Especificaciones
| Type | Penicillin G Sodium Salt |
| Physical Form | Powder |
| Color | White |
| Quantity | 5 g |
| Beilstein | 3834217 |
| Merck Index | 14,7094 |
| Solubility Information | Soluble in water (100mg/ml),alcohol,glycerol,DMSO (71mg/ml at 25°C),and ethanol (< 1mg/ml at 25°C). |
| Formula Weight | 356.37 |
| Chemical Name or Material | Penicillin G sodium salt |
RUO – Research Use Only
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.