missing translation for 'onlineSavingsMsg'
Learn More

Penicillin/Streptomycin/Neomycin Mixture, Fisher BioReagents

Product Code. 10063967
Change view
Click to view available options
Quantity:
50 mL
Unit Size:
50mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10063967 50 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10063967 Supplier Fisher BioReagents Supplier No. BP296150
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Interferes with synthesis of bacterial cell wall. Inhibits protein synthesis

  • An antibiotic mixture used to prevent bacterial contamination of cell cultures
  • Three combined antibiotics prove effective against gram-positive and gram-negative bacteria
  • Penicillin interferes with the synthesis of bacterial cell wall
  • Streptomycin and neomycin inhibit protein synthesis in bacteria by binding to the 30S subunit of bacterial ribosome
  • Intended for research purposes only
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Penicillin/Streptomycin/Neomycin Mixture
Type Penicillin/Streptomycin/Neomycin
Physical Form Liquid
CAS 1405-10-3, 3810-74-0, 69-57-8, 7647-14-5, 7732-18-5
Packaging Plastic Bottle
Sterile Filtered Pass Test
Quantity 50 mL
Merck Index 15, 6539
Synonym PSN Mixture
compliance-icons
Product Identifier
  • Penicillin/Streptomycin/Neomycin Mixture
Signal Word
  • Danger
Hazard Category
  • Reproductive toxicity Category 1B
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H360FD-May damage fertility. May damage the unborn child.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.