missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Penicillin-Streptomycin Solution (10,000 units ea.)

Product Code. 11558876
Change view
Click to view available options
Quantity:
20 x 100mL
Unit Size:
Pack of 20
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11558876 20 x 100mL Pack of 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11558876 Supplier Gibco™ Supplier No. 15140130
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Penicillin-Streptomycin Solution (10,000 units ea.)

The combination of the antibiotics penicillin and streptomycin are used to prevent bacterial contamination of cell cultures due to their effective action against gram-positive and gram-negative bacteria, respectively. Penicillin was originally purified from the fungi Penicillium and acts by interfering directly with the turnover of the bacteria cell wall and indirectly by triggering the release of enzymes that further alter the cell wall. Streptomycin was originally purified from Streptomyces griseus. Streptomycin acts by binding to the 30S subunit of the bacterial ribosome leading to inhibition of protein synthesis and death in susceptible bacteria. We offer a variety of antibiotics and antimycotics for cell culture applications.

This solution contains 10000 units of penicillin and 10000μg of streptomycin per ml.

Product Use
For Research Use Only: Not intended for animal or human diagnostic or therapeutic use.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage –5 to –20°C
Description Penicillin-Streptomycin Solution, 20 x 100mL, 100X, –5 to –20°C Storage Temperature, Antibiotic, Penicillin, Streptomycin Agent, Room Temperature/Frozen, Contains 10000 Units of Penicillin and 10000μg of Streptomycin PermL
Concentration 100 X
Form Liquid
Product Type Antibiotic
For Use With (Application) Prevention of cell culture contamination
Quantity 20 x 100mL

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.