missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pennsylvania Pesticide Mixture 333 10 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | Abamectin∣Acephate∣Acequinocyl∣Acetamiprid∣Aldicarb∣Azoxystrobin∣Bifenazate∣Bifenthrin∣Boscalid∣Captan∣Carbaryl∣Carbofuran∣Chlorantraniliprole∣Chlorfenapyr∣Chlorpyrifos∣Clofentezine∣Cyfluthrin∣Cypermethrin∣Daminozide∣Diazinon∣Dichlorvos∣Dimethoate∣Dimethomorph∣Ethoprophos∣Etofenprox∣Etoxazole∣Fenhexamid∣Fenoxycarb∣Fenpyroximate (E/Z Mix)∣Fipronil∣Flonicamid∣Fludioxonil∣Hexythiazox∣Imazalil∣Imidacloprid∣Kresoxim-methyl∣Malathion∣Metalaxyl∣Methiocarb∣Methomyl∣MGK 264∣Myclobutanil∣Naled∣Oxamyl∣Paclobutrazol∣Parathion-methyl∣Permethrin∣Phosmet∣Piperonyl butoxide∣Prallethrin∣Propiconazole∣Propoxur∣Pyridaben∣Spinetoram∣Spinosad∣Spiromesifen∣Spirotetramat∣Spiroxamine∣Tebuconazole∣Thiacloprid∣Thiamethoxam∣Trifloxystrobin |
| CAS | 71751-41-2, 30560-19-1, 57960-19-7, 135410-20-7, 116-06-3, 131860-33-8 |
| Analyte or Component Names | Abamectin∣Acephate∣Acequinocyl∣Acetamiprid∣Aldicarb∣Azoxystrobin∣Bifenazate∣Bifenthrin∣Boscalid∣Captan∣Carbaryl∣Carbofuran∣Chlorantraniliprole∣Chlorfenapyr∣Chlorpyrifos∣Clofentezine∣Cyfluthrin∣Cypermethrin∣Daminozide∣Diazinon∣Dichlorvos∣Dimethoate∣Dimethomorph∣Ethoprophos∣Etofenprox∣Etoxazole∣Fenhexamid∣Fenoxycarb∣Fenpyroximate (E/Z Mix)∣Fipronil∣Flonicamid∣Fludioxonil∣Hexythiazox∣Imazalil∣Imidacloprid∣Kresoxim-methyl∣Malathion∣Metalaxyl∣Methiocarb∣Methomyl∣MGK 264∣Myclobutanil∣Naled∣Oxamyl∣Paclobutrazol∣Parathion-methyl∣Permethrin∣Phosmet∣Piperonyl butoxide∣Prallethrin∣Propiconazole∣Propoxur∣Pyridaben∣Spinetoram∣Spinosad∣Spiromesifen∣Spirotetramat∣Spiroxamine∣Tebuconazole∣Thiacloprid∣Thiamethoxam∣Trifloxystrobin |
| Shipping Condition | Room Temperature |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Acetonitrile |
| Quantity | 1 mL |
| Concentration or Composition (by Analyte or Components) | Abamectin 10 μg/mL∣Acephate 10 μg/mL∣Acequinocyl 10 μg/mL∣Acetamiprid 10 μg/mL∣Aldicarb 10 μg/mL∣Azoxystrobin 10 μg/mL∣Bifenazate 10 μg/mL∣Bifenthrin 10 μg/mL∣Boscalid 10 μg/mL∣Captan 10 μg/mL∣Carbaryl 10 μg/mL∣Carbofuran 10 μg/mL∣Chlorantraniliprole 10 μg/mL∣Chlorfenapyr 10 μg/mL∣Chlorpyrifos 10 μg/mL∣Clofentezine 10 μg/mL∣Cyfluthrin 10 μg/mL∣Cypermethrin 10 μg/mL∣Daminozide 10 μg/mL∣Diazinon 10 μg/mL∣Dichlorvos 10 μg/mL∣Dimethoate 10 μg/mL∣Dimethomorph 10 μg/mL∣Ethoprophos 10 μg/mL∣Etofenprox 10 μg/mL∣Etoxazole 10 μg/mL∣Fenhexamid 10 μg/mL∣Fenoxycarb 10 μg/mL∣Fipronil 10 μg/mL∣Flonicamid 10 μg/mL∣Fludioxonil 10 μg/mL∣Hexythiazox 10 μg/mL∣Imazalil 10 μg/mL∣Imidacloprid 10 μg/mL∣Kresoxim-methyl 10 μg/mL∣Malathion 10 μg/mL∣Metalaxyl 10 μg/mL∣Methiocarb 10 μg/mL∣Methomyl 10 μg/mL∣MGK 264 10 μg/mL∣Myclobutanil 10 μg/mL∣Naled 10 μg/mL∣Oxamyl 10 μg/mL∣Paclobutrazol 10 μg/mL∣Parathion-methyl 10 μg/mL∣Permethrin 10 μg/mL∣Phosmet 10 μg/mL∣Piperonyl butoxide 10 μg/mL∣Prallethrin 10 μg/mL∣Propiconazole 10 μg/mL∣Propoxur 10 μg/mL∣Pyridaben 10 μg/mL∣Spinetoram 10 μg/mL∣Spinosad 10 μg/mL∣Spiromesifen 10 μg/mL∣Spirotetramat 10 μg/mL∣Spiroxamine 10 μg/mL∣Tebuconazole 10 μg/mL∣Thiacloprid 10 μg/mL∣Thiamethoxam 10 μg/mL∣Trifloxystrobin 10 μg/mL |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?