missing translation for 'onlineSavingsMsg'
Learn More

Pentacene, 98%

Product Code. 10406950 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mg
1 g
Packaging:
Glass bottle
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10406950 100 mg Glass bottle 100mg
10751414 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10406950 Supplier Thermo Scientific Acros Supplier No. 129791000
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 135-48-8 | C22H14 | 278.35 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 135-48-8
Molecular Formula C22H14
Molecular Weight (g/mol) 278.35
MDL Number MFCD00003710
InChI Key SLIUAWYAILUBJU-UHFFFAOYSA-N
Synonym benzo b naphthacene, lin-dibenzanthracene, lin-naphthoanthracene, 2,3:6,7-dibenzanthracene, unii-9fqu5ha0uy, 9fqu5ha0uy, pentacen-2-yl, pentacene, 2,7-dibenzanthracene
PubChem CID 8671
ChEBI CHEBI:33148
IUPAC Name pentacene
SMILES C1=CC2=CC3=CC4=CC5=CC=CC=C5C=C4C=C3C=C2C=C1

Specifications

Melting Point >300°C
Color Black or Blue-Purple
Infrared Spectrum Authentic
Assay Percent Range 97.5% min
Packaging Glass bottle
Quantity 100 mg
Merck Index 15, 7216
Solubility Information Solubility in water: insoluble. Other solubilities: slightly soluble in benzene,soluble in nitrobenzene
Formula Weight 278.34
Percent Purity 98%
Physical Form Fine Crystalline Powder
Chemical Name or Material Pentacene, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.