missing translation for 'onlineSavingsMsg'
Learn More

Pentachlorophenol 1000 μg/mL in Methanol, Dr. Ehrenstorfer

Product Code. 30313341
Change view
Click to view available options
Quantity:
5 x 1 mL
Unit Size:
5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30313341 5 x 1 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30313341 Supplier Dr. Ehrenstorfer Supplier No. DREYS09010008ME
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Pentachlorophenol
CAS 87-86-5
Analyte or Component Names Pentachlorophenol
Shipping Condition Room Temperature
Molecular Formula C6 H Cl5 O
Concentration 1000 μg/mL
Certifications/Compliance ISO 17034
Synonym Phenol, 2,3,4,5,6-pentachloro-, Phenol, pentachloro- (8CI,9CI), 2,3,4,5,6-Pentachlorophenol, 1-Hydroxy-2,3,4,5,6-pentachlorobenzene, 1-Hydroxypentachlorobenzene, Chlon, Chlorophenasic acid, Dowicide 7, Dowicide EC 7, Dura Treet II, EP 30, EP 30 (pesticide), Fungifen, Grundier Arbezol, Lauxtol, Liroprem, NSC 263497, PCP, PCP (pesticide), PKhF, Penchlorol, Penta, Pentachlorophenol, Penton 70, Pentor 70, Perchlorophenol, Permasan, Pol Nu, Pole topper, Pole topper fluid, Preventol P, Santophen 20, Satophen, Witophen P, Woodtreat A
InChI Formula InChI=1S/C6HCl5O/c7-1-2(8)4(10)6(12)5(11)3(1)9/h12H
SMILES Oc1c(Cl)c(Cl)c(Cl)c(Cl)c1Cl
IUPAC Name 2,3,4,5,6-pentachlorophenol
Solvent or Matrix Methanol
Industry Type Food Testing
Molecular Weight (g/mol) 266.34
Quantity 5 x 1 mL
Formula Weight 263.847 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.