missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pentadecafluorooctanoic acid, 96%
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 53.0°C to 60.0°C |
| Color | White |
| Boiling Point | 189.0°C to 192.0°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 96% |
| Packaging | Glass bottle |
| Linear Formula | CF3(CF2)6CO2H |
| Quantity | 100 g |
| Merck Index | 15, 7272 |
| Solubility Information | Solubility in water: 3.4g/L (20°C). Other solubilities: soluble in ether and ethanol |
| Show More |
Safety and Handling
Product Identifier
- Pentadecafluorooctanoic acid
- Danger
- Acute toxicity Category 4
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 1
- Reproductive toxicity on or via lactation
- Reproductive toxicity Category 1B
- Specific target organ toxicity Category 1
- H318-Causes serious eye damage.
- H372-Causes damage to organs.
- H360D-May damage the unborn child.
- H351-Suspected of causing cancer.
- H332-Harmful if inhaled.
- H362-May cause harm to breast-fed children.
- H302-Harmful if swallowed.
- P263-Avoid contact during pregnancy/while nursing.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction