missing translation for 'onlineSavingsMsg'
Learn More

Pentafluorobenzaldehyde, 98%

Product Code. 11429160
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11429160 100 g 100g
11409160 5 g 5g
11419160 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11429160 Supplier Thermo Scientific Alfa Aesar Supplier No. A15490.22
 Restricted Item
Estimated Shipment: 22-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 653-37-2 | C7HF5O | 196.076 g/mol

Pentafluorobenzaldehyde was used as derivatization reagent in a study to develop a sensitive method for the determination of primary amines in sewage sludge by headspace solid-phase micro extraction and gas chromatography-tandem mass spectrometry. It was used in the synthesis of novel, high molecular weight fluorinated aromatic polymers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pentafluorobenzaldehyde was used as derivatization reagent in a study to develop a sensitive method for the determination of primary amines in sewage sludge by headspace solid-phase micro extraction and gas chromatography-tandem mass spectrometry. It was used in the synthesis of novel, high molecular weight fluorinated aromatic polymers.

Solubility
Sparingly Soluble in water (0.027 g/L at 25°C).

Notes
Store in a cool, dry conditions in a well sealed container. Incompatible with oxidizing agent, air, bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 653-37-2
Molecular Formula C7HF5O
Molecular Weight (g/mol) 196.076
MDL Number MFCD00003303
InChI Key QJXCFMJTJYCLFG-UHFFFAOYSA-N
Synonym pentafluorobenzaldehyde, perfluorobenzaldehyde, benzaldehyde, pentafluoro, benzaldehyde, 2,3,4,5,6-pentafluoro, unii-s9yie3jz4k, pentafluor-benzaldehyd, s9yie3jz4k, pubchem4253, pentafluoro-benzaldehyde, acmc-209nqh
PubChem CID 69558
IUPAC Name 2,3,4,5,6-pentafluorobenzaldehyde
SMILES C(=O)C1=C(C(=C(C(=C1F)F)F)F)F

Specifications

Melting Point 24°C to 27°C
Density 1.63
Boiling Point 164°C to 166°C
Flash Point 77°C (170°F)
Refractive Index 1.451
Quantity 100 g
UN Number UN2811
Beilstein 1876550
Sensitivity Air Sensitive
Solubility Information Sparingly Soluble in water (0.027g/L at 25°C).
Formula Weight 196.07
Percent Purity ≥98%
Chemical Name or Material Pentafluorobenzaldehyde
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.