missing translation for 'onlineSavingsMsg'
Learn More

Pentafluorobenzoic acid, 98+%

Product Code. 11479553
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11479553

Brand: Thermo Scientific Alfa Aesar A12396.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 602-94-8 | C7HF5O2 | 212.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 602-94-8
Molecular Formula C7HF5O2
Molecular Weight (g/mol) 212.08
MDL Number MFCD00002406
InChI Key YZERDTREOUSUHF-UHFFFAOYSA-N
Synonym pentafluorobenzoic acid, perfluorobenzoic acid, benzoic acid, pentafluoro, benzoic acid, 2,3,4,5,6-pentafluoro, 2,3,4,5,6-pentafluorobenzoicacid, pubchem2320, acmc-1b1wz, 4-09-00-00956 beilstein handbook reference, ksc355e3b, rarechem al bo 0017
PubChem CID 11770
ChEBI CHEBI:46796
IUPAC Name 2,3,4,5,6-pentafluorobenzoic acid
SMILES OC(=O)C1=C(F)C(F)=C(F)C(F)=C1F

Specifications

Melting Point 100°C to 104°C
Boiling Point 219°C to 221°C
Quantity 5 g
Beilstein 2054395
Formula Weight 212.07
Percent Purity 99%
Chemical Name or Material Pentafluorobenzoic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.