missing translation for 'onlineSavingsMsg'
Learn More

Pentafluorobenzonitrile, 99%

Product Code. 15500524 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15500524 5 g 5g
15510524 25 g 25g
15520524 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15500524 Supplier Thermo Scientific Acros Supplier No. 188340050
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 773-82-0 | C7F5N | 193.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 773-82-0
Molecular Formula C7F5N
Molecular Weight (g/mol) 193.07
InChI Key YXWJGZQOGXGSSC-UHFFFAOYSA-N
Synonym pentafluorobenzonitrile, perfluorobenzonitrile, benzonitrile, pentafluoro, pentafluorocyanobenzene, benzonitrile, 2,3,4,5,6-pentafluoro, 2,3,4,5,6-pentafluoro benzonitrile, 2,3,4,5,6-pentafluorobenzenecarbonitrile, cyanopentafluorobenzene, pubchem2319, acmc-209p9k
PubChem CID 69882
IUPAC Name 2,3,4,5,6-pentafluorobenzonitrile
SMILES C(#N)C1=C(C(=C(C(=C1F)F)F)F)F

Specifications

Melting Point 1.0°C to 3.0°C
Density 1.5300g/mL
Boiling Point 161.0°C to 162.0°C
Flash Point 29°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Linear Formula C6F5CN
Refractive Index 1.4415 to 1.4435
Quantity 5 g
Specific Gravity 1.53
Solubility Information Solubility in water: insoluble.
Formula Weight 193.07
Percent Purity 99%
Chemical Name or Material Pentafluorobenzonitrile
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Pentafluorobenzonitrile
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.