missing translation for 'onlineSavingsMsg'
Learn More

Pentafluorobenzonitrile, 99%

Product Code. 15520524 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15520524 100 g 100g
15500524 5 g 5g
15510524 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15520524 Supplier Thermo Scientific Acros Supplier No. 188341000
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 773-82-0 | C7F5N | 193.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 773-82-0
Molecular Formula C7F5N
Molecular Weight (g/mol) 193.07
InChI Key YXWJGZQOGXGSSC-UHFFFAOYSA-N
Synonym pentafluorobenzonitrile, perfluorobenzonitrile, benzonitrile, pentafluoro, pentafluorocyanobenzene, benzonitrile, 2,3,4,5,6-pentafluoro, 2,3,4,5,6-pentafluoro benzonitrile, 2,3,4,5,6-pentafluorobenzenecarbonitrile, cyanopentafluorobenzene, pubchem2319, acmc-209p9k
PubChem CID 69882
IUPAC Name 2,3,4,5,6-pentafluorobenzonitrile
SMILES C(#N)C1=C(C(=C(C(=C1F)F)F)F)F

Especificaciones

Melting Point 1.0°C to 3.0°C
Density 1.5300g/mL
Boiling Point 161.0°C to 162.0°C
Flash Point 29°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Linear Formula C6F5CN
Refractive Index 1.4415 to 1.4435
Quantity 100 g
Specific Gravity 1.53
Solubility Information Solubility in water: insoluble.
Formula Weight 193.07
Percent Purity 99%
Chemical Name or Material Pentafluorobenzonitrile
Mostrar más Mostrar menos
compliance-icons compliance-icons
Nombre del producto
  • Pentafluorobenzonitrile
Palabra de advertencia
  • Danger
Clasificación
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Indicaciones de peligro
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H226-Flammable liquid and vapour.
Consejos de prudencia
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.