missing translation for 'onlineSavingsMsg'
Learn More

Pentafluoroiodobenzene, 97%

Product Code. 11464044
Change view
Click to view available options
Quantity:
5 g
10 g
50 g
Unit Size:
10g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11464044 10 g 10g
11428690 5 g 5g
11474044 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11464044 Supplier Thermo Scientific Alfa Aesar Supplier No. A12000.09
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 827-15-6 | C6F5I | 293.962 g/mol

Iodopentafluorobenzene was used to study the formation of radical ions of iodopentafluorobenzene in aqueous solution. It was used as solvent in a study to determine singlet oxygen lifetimes from phosphorescence decays in halogen substituted perfluorinated solvents by infrared emission spectrometery. It has potential applications in plasma processing industry and in preparation of catalysts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Iodopentafluorobenzene was used to study the formation of radical ions of iodopentafluorobenzene in aqueous solution. It was used as solvent in a study to determine singlet oxygen lifetimes from phosphorescence decays in halogen substituted perfluorinated solvents by infrared emission spectrometery. It has potential applications in plasma processing industry and in preparation of catalysts.

Solubility
Insoluble in water.

Notes
Light sensitive. Store at room temperature. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 827-15-6
Molecular Formula C6F5I
Molecular Weight (g/mol) 293.962
MDL Number MFCD00001032
InChI Key OPYHNLNYCRZOGY-UHFFFAOYSA-N
Synonym iodopentafluorobenzene, pentafluoroiodobenzene, benzene, pentafluoroiodo, 2,3,4,5,6-pentafluoroiodobenzene, iodoperfluorobenzene, pentafluorophenyl iodide, 1,2,3,4,5-pentafluoro-6-iodo-benzene, perfluoroiodobenzene, ibf, pubchem19982
PubChem CID 70008
IUPAC Name 1,2,3,4,5-pentafluoro-6-iodobenzene
SMILES C1(=C(C(=C(C(=C1F)F)I)F)F)F

Specifications

Melting Point -29°C
Density 2.212
Boiling Point 161°C to 163°C
Refractive Index 1.496
Quantity 10 g
Beilstein 2051549
Sensitivity Light sensitive
Solubility Information Insoluble in water.
Formula Weight 293.96
Percent Purity 97%
Chemical Name or Material Pentafluoroiodobenzene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.