missing translation for 'onlineSavingsMsg'
Learn More

Pentafluorophenyl 2-morpholino-1,3-thiazole-4-carboxylate, 97%, Thermo Scientific™

Product Code. 10595954
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10595954 250 mg Amber Glass Bottle 250mg
10367314 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10595954 Supplier Thermo Scientific Maybridge Supplier No. CC62826CB
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 921939-02-8
Molecular Formula C14H9F5N2O3S
Molecular Weight (g/mol) 380.29
MDL Number MFCD09817538
InChI Key NABJYXXXUIZUOT-UHFFFAOYSA-N
Synonym pentafluorophenyl 2-morpholin-4-yl-1,3-thiazole-4-carboxylate, pentafluorophenyl 2-morpholino-1,3-thiazole-4-carboxylate, 2,3,4,5,6-pentafluorophenyl 2-morpholin-4-yl-1,3-thiazole-4-carboxylate, 2,3,4,5,6-pentakis fluoranyl phenyl 2-morpholin-4-yl-1,3-thiazole-4-carboxylate, 2-4-morpholinyl-4-thiazolecarboxylic acid 2,3,4,5,6-pentafluorophenyl ester
PubChem CID 24229699
IUPAC Name (2,3,4,5,6-pentafluorophenyl) 2-morpholin-4-yl-1,3-thiazole-4-carboxylate
SMILES FC1=C(F)C(F)=C(OC(=O)C2=CSC(=N2)N2CCOCC2)C(F)=C1F

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 380.29
Percent Purity 97%
Chemical Name or Material Pentafluorophenyl 2-morpholino-1, 3-thiazole-4-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.