missing translation for 'onlineSavingsMsg'
Learn More

Pentafluorophenyl 2-(morpholinosulfonyl)benzoate, 97%, Thermo Scientific™

Product Code. 10265464
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10265464 1 g Amber Glass Bottle 1g
10769215 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10265464 Supplier Thermo Scientific Maybridge Supplier No. CC69626DA
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 950603-27-7
Molecular Formula C17H12F5NO5S
Molecular Weight (g/mol) 437.34
MDL Number MFCD09879986
InChI Key FDXYCKHOIXBDOA-UHFFFAOYSA-N
Synonym pentafluorophenyl 2-morpholinosulfonyl benzoate, pentafluorophenyl 2-morpholine-4-sulfonyl benzoate, 2,3,4,5,6-pentafluorophenyl 2-morpholine-4-sulfonyl benzoate, pentafluorophenyl 2-morpholin-4-ylsulphonyl benzoate, 2,3,4,5,6-pentafluorophenyl 2-morpholinosulfonylbenzoate, 2,3,4,5,6-pentafluorophenyl 2-4-morpholinylsulfonyl benzoate, 2,3,4,5,6-pentakis fluoranyl phenyl 2-morpholin-4-ylsulfonylbenzoate, 2-4-morpholinylsulfonyl benzoic acid 2,3,4,5,6-pentafluorophenyl ester
PubChem CID 24229773
IUPAC Name (2,3,4,5,6-pentafluorophenyl) 2-morpholin-4-ylsulfonylbenzoate
SMILES FC1=C(F)C(F)=C(OC(=O)C2=CC=CC=C2S(=O)(=O)N2CCOCC2)C(F)=C1F

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 437.34
Percent Purity 97%
Chemical Name or Material Pentafluorophenyl 2-(morpholinosulfonyl)benzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.