missing translation for 'onlineSavingsMsg'
Learn More

Pentafluorophenyl 2-pyrid-3-yl-1,3-thiazole-4-carboxylate, 97%, Thermo Scientific™

Product Code. 11330577
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11330577 250 mg 250mg
10306544 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11330577 Supplier Thermo Scientific Maybridge Supplier No. CC13726CB
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 906352-58-7
Molecular Formula C15H5F5N2O2S
Molecular Weight (g/mol) 372.27
MDL Number MFCD09025821
InChI Key LHRLGPDYSCUADI-UHFFFAOYSA-N
Synonym pentafluorophenyl 2-pyrid-3-yl-1,3-thiazole-4-carboxylate, pentafluorophenyl 2-pyridin-3-yl-1,3-thiazole-4-carboxylate, 2,3,4,5,6-pentafluorophenyl 2-pyridin-3-yl-1,3-thiazole-4-carboxylate, perfluorophenyl 2-pyridin-3-yl thiazole-4-carboxylate, 2,3,4,5,6-pentakis fluoranyl phenyl 2-pyridin-3-yl-1,3-thiazole-4-carboxylate, 2-3-pyridinyl-4-thiazolecarboxylic acid 2,3,4,5,6-pentafluorophenyl ester
PubChem CID 18525705
SMILES FC1=C(F)C(F)=C(OC(=O)C2=CSC(=N2)C2=CN=CC=C2)C(F)=C1F

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 250 mg
IUPAC Name 2,3,4,5,6-pentafluorophenyl 2-(pyridin-3-yl)-1,3-thiazole-4-carboxylate
Formula Weight 372.27
Percent Purity 97%
Chemical Name or Material Pentafluorophenyl 2-pyrid-3-yl-1, 3-thiazole-4-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.