missing translation for 'onlineSavingsMsg'
Learn More

Pentafluorophenyl 3-(pyrid-2-yloxy)benzoate, 97%, Thermo Scientific™

Product Code. 10707675
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10707675 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10707675 Supplier Thermo Scientific Maybridge Supplier No. CC50226DA
 Restricted Item
Estimated Shipment: 05-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 921938-61-6
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C18H8F5NO3
MDL Number MFCD09064956
Quantity 1 g
Synonym pentafluorophenyl 3-pyrid-2-yloxy benzoate, pentafluorophenyl 3-pyridin-2-yloxy benzoate, pentafluorophenyl 3-pyridin-2-yl oxy benzoate, 2,3,4,5,6-pentafluorophenyl 3-pyridin-2-yloxy benzoate, 2,3,4,5,6-pentakis fluoranyl phenyl 3-pyridin-2-yloxybenzoate, 3-2-pyridinyloxy benzoic acid 2,3,4,5,6-pentafluorophenyl ester
InChI Key LKCOLTBNDUOLEF-UHFFFAOYSA-N
SMILES C1=CC=NC(=C1)OC2=CC=CC(=C2)C(=O)OC3=C(C(=C(C(=C3F)F)F)F)F
IUPAC Name (2,3,4,5,6-pentafluorophenyl) 3-pyridin-2-yloxybenzoate
Molecular Weight (g/mol) 381.258
PubChem CID 24229506
Formula Weight 381.26
Percent Purity 97%
Chemical Name or Material Pentafluorophenyl 3-(pyrid-2-yloxy)benzoate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.