missing translation for 'onlineSavingsMsg'
Learn More

Pentafluorophenyl 6-thien-2-ylnicotinate, 97%, Thermo Scientific™

Product Code. 10367534
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10367534 250 mg Amber Glass Bottle 250mg
10472284 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10367534 Supplier Thermo Scientific Maybridge Supplier No. CC54826CB
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 926921-59-7
Molecular Formula C16H6F5NO2S
Molecular Weight (g/mol) 371.28
MDL Number MFCD09702374
InChI Key UYAXHNWJTLZXTM-UHFFFAOYSA-N
Synonym pentafluorophenyl 2-thien-2-ylpyridine-5-carboxylate, pentafluorophenyl 6-thien-2-ylnicotinate, pentafluorophenyl 6-thiophen-2-yl pyridine-3-carboxylate, 2,3,4,5,6-pentafluorophenyl 6-thiophen-2-yl pyridine-3-carboxylate, pentafluorophenyl 6-thiophen-2-ylnicotinate, 2,3,4,5,6-pentakis fluoranyl phenyl 6-thiophen-2-ylpyridine-3-carboxylate, 6-thiophen-2-yl-3-pyridinecarboxylic acid 2,3,4,5,6-pentafluorophenyl ester
PubChem CID 24229543
IUPAC Name (2,3,4,5,6-pentafluorophenyl) 6-thiophen-2-ylpyridine-3-carboxylate
SMILES FC1=C(F)C(F)=C(OC(=O)C2=CN=C(C=C2)C2=CC=CS2)C(F)=C1F

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 371.28
Percent Purity 97%
Chemical Name or Material Pentafluorophenyl 6-thien-2-ylnicotinate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.