missing translation for 'onlineSavingsMsg'
Learn More

Pentafluorophenyl nicotinate, 97%, Thermo Scientific™

Product Code. 10396794
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10396794 1 g Amber Glass Bottle 1g
10143214 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10396794 Supplier Thermo Scientific Maybridge Supplier No. CC04126DA
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 848347-44-4
Molecular Formula C12H4F5NO2
Molecular Weight (g/mol) 289.161
MDL Number MFCD09064938
InChI Key AXHLJDBUUFUDCF-UHFFFAOYSA-N
Synonym pentafluorophenyl nicotinate, pentafluorophenyl pyridine-3-carboxylate, 2,3,4,5,6-pentakis fluoranyl phenyl pyridine-3-carboxylate, 3-pyridinecarboxylic acid 2,3,4,5,6-pentafluorophenyl ester
PubChem CID 23237920
IUPAC Name (2,3,4,5,6-pentafluorophenyl) pyridine-3-carboxylate
SMILES C1=CC(=CN=C1)C(=O)OC2=C(C(=C(C(=C2F)F)F)F)F

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 289.16
Percent Purity 97%
Chemical Name or Material Pentafluorophenyl nicotinate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.