missing translation for 'onlineSavingsMsg'
Learn More

Pentafluorophenyl pyridine-2-carboxylate, 95%, Thermo Scientific™

Product Code. 10777855
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10777855 1 g Amber Glass Bottle 1g
10041714 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10777855 Supplier Thermo Scientific Maybridge Supplier No. CC04026DA
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 188837-53-8
Molecular Formula C12H4F5NO2
Molecular Weight (g/mol) 289.16
MDL Number MFCD02916457
InChI Key KFJSLIBBYMXLTG-UHFFFAOYSA-N
Synonym pentafluorophenyl pyridine-2-carboxylate, pentafluorophenyl picolinate, 2-pyridinecarboxylicacid, 2,3,4,5,6-pentafluorophenyl ester, 2,3,4,5,6-pentafluorophenyl pyridine-2-carboxylate, picolinic acid pentafluorophenyl ester, 2-pentafluorophenoxy carbonyl pyridine, 2,3,4,5,6-pentakis fluoranyl phenyl pyridine-2-carboxylate, 2-pyridinecarboxylic acid 2,3,4,5,6-pentafluorophenyl ester
PubChem CID 2754830
IUPAC Name (2,3,4,5,6-pentafluorophenyl) pyridine-2-carboxylate
SMILES FC1=C(F)C(F)=C(OC(=O)C2=CC=CC=N2)C(F)=C1F

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 289.16
Percent Purity 95%
Chemical Name or Material Pentafluorophenyl pyridine-2-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.