missing translation for 'onlineSavingsMsg'
Learn More

Pentafluorophenylhydrazine, 97%, Thermo Scientific Chemicals

Product Code. 11440893
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11440893 100 g 100g
11420893 5 g 5g
11430893 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11440893 Supplier Thermo Scientific Alfa Aesar Supplier No. A13302.22
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 828-73-9 | Molecular Formula: C6H3F5N2 | Molecular Weight: 198.096 g/mol

Pentafluorophenylhydrazine used to determine the purity of N-tert-butyl-N-[1-diethylphosphono-(2,2-dimethylpropyl)]nitroxide (DEPN) using 1H NMR spectroscopy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pentafluorophenylhydrazine used to determine the purity of N-tert-butyl-N-[1-diethylphosphono-(2,2-dimethylpropyl)]nitroxide (DEPN) using 1H NMR spectroscopy.

Solubility
Soluble in methanol.

Notes
Store at room temperature. Incompatible with oxidizing agents. Ensure adequate ventilation.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 828-73-9
Molecular Formula C6H3F5N2
Molecular Weight (g/mol) 198.096
MDL Number MFCD00007574
InChI Key BYCUWCJUPSUFBX-UHFFFAOYSA-N
Synonym pentafluorophenylhydrazine, pentafluorophenyl hydrazine, perfluorophenyl hydrazine, hydrazine, pentafluorophenyl, 2,3,4,5,6-pentafluorophenyl hydrazine, 1-2,3,4,5,6-pentafluorophenyl hydrazine, pubchem3316, pentafluorophenyl-hydrazine
PubChem CID 13236
IUPAC Name (2,3,4,5,6-pentafluorophenyl)hydrazine
SMILES C1(=C(C(=C(C(=C1F)F)F)F)F)NN

Specifications

Melting Point 73°C to 78°C
Quantity 100 g
Beilstein 747800
Sensitivity Air Sensitive
Solubility Information Soluble in methanol.
Formula Weight 198.09
Percent Purity 97%
Chemical Name or Material Pentafluorophenylhydrazine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.