missing translation for 'onlineSavingsMsg'
Learn More

Pentafluorophenylmagnesium bromide, 0.5M solution in diethyl ether, AcroSeal™

Product Code. 11945991 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11945991 100 mL AcroSeal™ Glass bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11945991 Supplier Thermo Scientific Acros Supplier No. 438741000
 Restricted Item
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 879-05-0 | C6BrF5Mg | 271.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 879-05-0, 60-29-7
Density 0.8600g/mL
Flash Point −40°C
Packaging AcroSeal™ Glass bottle
Molecular Formula C6BrF5Mg
Linear Formula F5C6MgBr
MDL Number MFCD00015720
Quantity 100 mL
Specific Gravity 0.86
Synonym pentafluorophenylmagnesium bromide, magnesium, bromo pentafluorophenyl, pentafluorophenylmagnesiumbromide, c6f5mgbr, pentafluorophenyl magnesiumbromide, axqvmjidndrsfm-uhfffaoysa-m, pentafluorophenyl magnesium bromide, pentafluorophenylmagnesium bromide solution, bromo 2,3,4,5,6-pentafluorophenyl magnesium, pentafluorophenylmagnesium bromide solution, 0.5 m in diethyl ether, grignard reagent
InChI Key AXQVMJIDNDRSFM-UHFFFAOYSA-M
SMILES FC1=C(F)C(F)=C([Mg]Br)C(F)=C1F
IUPAC Name magnesium;1,2,3,4,5-pentafluorobenzene-6-ide;bromide
Molecular Weight (g/mol) 271.27
PubChem CID 3319152
Formula Weight 271.27
Chemical Name or Material Pentafluorophenylmagnesium bromide
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Pentafluorophenylmagnesium bromide, 0.5M solution in diethyl ether
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H336-May cause drowsiness or dizziness.
  • H302-Harmful if swallowed.
  • H224-Extremely flammable liquid and vapour.
  • EUH014-Reacts violently with water.
  • EUH019-May form explosive peroxides.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands and any thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.