missing translation for 'onlineSavingsMsg'
Learn More

Pentafluoropropionic acid, 97%

Product Code. 10239850 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Plastic bottle
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10239850 50 g Plastic bottle 50g
10035870 10 g Plastic bottle 10g
15790869 250 g Plastic bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10239850 Supplier Thermo Scientific Acros Supplier No. 416920500
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 422-64-0 | C3HF5O2 | 164.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 422-64-0
Molecular Formula C3AgF5O2
Molecular Weight (g/mol) 270.89
MDL Number MFCD00004170
InChI Key XAYJXAUUXJTOSI-UHFFFAOYSA-M
Synonym pentafluoropropionic acid, perfluoropropionic acid, pentafluoropropanoic acid, propanoic acid, pentafluoro, propionic acid, pentafluoro, 2,2,3,3,3-pentafluoropropionic acid, ipc-pffa-3, perfluoropropanoic acid, propanoic acid, 2,2,3,3,3-pentafluoro, perfluoropropionicacid
PubChem CID 62356
SMILES [Ag+].[O-]C(=O)C(F)(F)C(F)(F)F

Specifications

Color Colorless to Brown
Density 1.5760g/mL
Boiling Point 96°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Plastic bottle
Linear Formula CF3CF2CO2H
Refractive Index 1.2830 to 1.286
Quantity 50 g
Specific Gravity 1.576
IUPAC Name 2,2,3,3,3-pentafluoropropanoic acid
Formula Weight 164.03
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Pentafluoropropionic acid
Show More Show Less
compliance-icons
Product Identifier
  • Pentafluoropropionic acid
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.