missing translation for 'onlineSavingsMsg'
Learn More

Pentafluorothiophenol, 97%

Product Code. 11416026
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11416026 5 g 5g
11426026 25 g 25g
11436026 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11416026 Supplier Thermo Scientific Alfa Aesar Supplier No. B22323.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 771-62-0 | C6HF5S | 200.13 g/mol

Pentafluorothiophenol is used as a fluorinated building block

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pentafluorothiophenol is used as a fluorinated building block

Solubility
Insoluble in water.

Notes
Air Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 771-62-0
Molecular Formula C6HF5S
Molecular Weight (g/mol) 200.13
MDL Number MFCD00004828
InChI Key UVAMFBJPMUMURT-UHFFFAOYSA-N
Synonym pentafluorothiophenol, pentafluorobenzenethiol, benzenethiol, pentafluoro, pentafluorophenyl mercaptan, 2,3,4,5,6-pentafluorothiophenol, 2,3,4,5,6-pentafluorobenzene-1-thiol, pentafluorobenzene thiol, pentafluorobenzene-1-thiol, 2,3,4,5,6-pentafluorothiphenol
PubChem CID 13042
IUPAC Name 2,3,4,5,6-pentafluorobenzenethiol
SMILES FC1=C(F)C(F)=C(S)C(F)=C1F

Specifications

Melting Point -24°C
Density 1.572
Boiling Point 143°C
Flash Point 51°C (123°F)
Odor Stench
Refractive Index 1.465
Quantity 5 g
UN Number UN2920
Beilstein 1876292
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 200.13
Percent Purity 97%
Chemical Name or Material Pentafluorothiophenol
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Pentafluorothiophenol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Pentafluorothiopheno

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.