missing translation for 'onlineSavingsMsg'
Learn More

Pentafluorothiophenol, 97%

Product Code. 10636332 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 mL
5 mL
25 mL
Packaging:
Glass bottle
Unit Size:
1mL
25mL
5mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10636332 5 mL Glass bottle 5mL
10254940 1 mL Glass bottle 1mL
10299393 25 mL Glass bottle 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10636332 Supplier Thermo Scientific Acros Supplier No. 129920050
 Restricted Item
Estimated Shipment: 18-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 771-62-0 | C6HF5S | 200.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 771-62-0
Molecular Formula C6HF5S
Molecular Weight (g/mol) 200.13
MDL Number MFCD00004828
InChI Key UVAMFBJPMUMURT-UHFFFAOYSA-N
Synonym pentafluorothiophenol, pentafluorobenzenethiol, benzenethiol, pentafluoro, pentafluorophenyl mercaptan, 2,3,4,5,6-pentafluorothiophenol, 2,3,4,5,6-pentafluorobenzene-1-thiol, pentafluorobenzene thiol, pentafluorobenzene-1-thiol, 2,3,4,5,6-pentafluorothiphenol
PubChem CID 13042
IUPAC Name 2,3,4,5,6-pentafluorobenzenethiol
SMILES FC1=C(F)C(F)=C(S)C(F)=C1F

Specifications

Melting Point -24°C
Color Colorless to Yellow
Density 1.5000g/mL
Boiling Point 143°C
Flash Point 51°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula C6F5SH
Refractive Index 1.4630 to 1.465
Quantity 5 mL
Specific Gravity 1.5
Formula Weight 200.13
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Pentafluorothiophenol
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Pentafluorothiophenol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.