missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pentraxin 3/TSG-14 Antibody (SR1716), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-21983-100ul
This item is not returnable.
View return policy
Beschreibung
Pentraxin 3/TSG-14 Monoclonal antibody specifically detects Pentraxin 3/TSG-14 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry
Spezifikation
| Pentraxin 3/TSG-14 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, Flow Cytometry | |
| SR1716 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:50 | |
| alpha-induced protein 5, pentaxin-related gene, rapidly induced by IL-1 beta, tumor necrosis factor, Pentaxin-related protein PTX3, pentraxin 3, long, pentraxin-3, pentraxin-related gene, rapidly induced by IL-1 beta, pentraxin-related protein PTX3, TNF alpha-induced protein 5, TNFAIP5, TSG14, TSG-14pentaxin-related gene, rapidly induced by IL-1 beta, Tumor necrosis factor alpha-induced protein 5, tumor necrosis factor, alpha-induced protein 5, Tumor necrosis factor-inducible gene 14 protein, tumor necrosis factor-inducible protein TSG-14 | |
| A synthesized peptide derived from human Pentraxin 3/TSG-14 (Uniprot #: P26022) | |
| 100 μg | |
| Immunology | |
| 5806 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur