missing translation for 'onlineSavingsMsg'
Learn More

Pepstatin, MedChemExpress

Product Code. 16479318
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16479318 10 mg 10mg
16474957 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16479318 Supplier Medchem Express Supplier No. HYP0018
 Restricted Item
Estimated Shipment: 14-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Pepstatin (Pepstatin A) is a specific aspartic protease inhibitor produced by actinomycetes, with IC50s of 4.5 nM, 6.2 nM, 150 nM, 290 nM, 520 nM and 260 nM for hemoglobin-pepsin, hemoglobin-proctase, casein-pepsin, casein-proctase, casein-acid protease and hemoglobin-acid protease, respectively. Pepstatin Ammonium also inhibits HIV protease.

Pepstatin (Pepstatin A) is a specific aspartic protease inhibitor produced by actinomycetes, with IC50s of 4.5 nM, 6.2 nM, 150 nM, 290 nM, 520 nM and 260 nM for hemoglobin-pepsin, hemoglobin-proctase, casein-pepsin, casein-proctase, casein-acid protease and hemoglobin-acid protease, respectively. Pepstatin Ammonium also inhibits HIV protease.

Specifications

Chemical Name or Material Pepstatin
Physical Form Solid
CAS 26305-03-3
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C34H63N5O9
For Use With (Application) COVID-19-anti-virus
Synonym Pepstatin A
Solubility Information DMSO : 41.67 mg/mL (60.75 mM; Need ultrasonic)
SMILES CC(C[C@H](NC([C@@H](NC([C@@H](NC(CC(C)C)=O)C(C)C)=O)C(C)C)=O)[C@@H](O)CC(N[C@@H](C)C(N[C@@H](CC(C)C)[C@@H](O)CC(O)=O)=O)=O)C
Molecular Weight (g/mol) 685.89
Formula Weight 685.89
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.