missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ PepSwift™ Monolithic Capillary HPLC Columns

Product Code. 11302023 Shop All Thermo Scientific Products
Change view
Click to view available options
Length (Metric):
5 mm
50 mm
250 mm
Diameter (Metric):
0.1 mm
0.2 mm
Unit Size:
Each
Pack of 2
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Diameter (Metric) unitSize
11302023 250 mm 0.1 mm Each
11322023 5 mm 0.2 mm Pack of 2
11312023 50 mm 0.2 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11302023 Supplier Thermo Scientific™ Supplier No. 164543

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Use for fast, high-resolution LC/MS separations in bottom-up protein identification, biomarker discovery, and systems biology applications.

For fast, high-resolution LC/MS separations in bottom-up protein identification, biomarker discovery, and systems biology applications, use Thermo Scientific™ PepSwift™ Monolithic Capillary LC Columns. Based on a polystyrene divinylbenzene copolymer, the monolithic structure offers a high-quality alternative to traditional microparticulate sorbents, providing important advantages to the chromatographic separation. High-sensitivity proteomics and biotech applications are easily performed using these columns.

Benefit from:

  • High-speed peptide and protein separations (<15 min)
  • High sensitivity for LC/MS
  • High column-to-column reproducibility
  • Wide range of column I.D. and lengths available
  • Superior lifetime
  • Easy column installation using Thermo Scientific™ Dionex™ nanoViper™ Fingertight Fittings
  • Precolumns available to preconcentrate and desalt samples

You can also use Thermo Scientific™ PepSwift™ Trap Columns for preconcentration and desalting of samples consisting of peptides and proteins without negatively impacting the chromatographic performance or recovery of the compounds. Change the selectivity of the separation or improve the trapping efficiency by employing various ion-pairing agents in the loading solvent and/or mobile phases. The trap columns are used with Thermo Scientific™ PepSwift™ and ProSwift™ Monolithic Columns, which are for fast, high-resolution LC/MS separations in protein identification, biomarker discovery, and systems biology applications.

TRUSTED_SUSTAINABILITY

Specifications

Column Format Analytical Column, NanoViper
Diameter (Metric) 0.1 mm
Length (Metric) 250 mm
Max. Pressure 4350 psi (300 bar)
Particle Size Monolith
pH 1 to 10
Pore Size Monolith
Temperature 80°C
Stationary Phase PSDVB (Polystyrene Divinylbenzene)
Product Line nanoLC
Packing Material Monolith Polymer
Column Type Reversed Phase
Phase Reversed Phase
Material Polystyrene/Divinylbenzene
Type Analytical Column, NanoViper
Show More Show Less

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.