Learn More
Thermo Scientific™ PepSwift™ Monolithic Capillary HPLC Columns
Use for fast, high-resolution LC/MS separations in bottom-up protein identification, biomarker discovery, and systems biology applications.
901.68 € - 1444.32 €
Specifications
Max. Pressure | 4350 psi (300 bar) |
---|---|
Particle Size | Monolith |
pH | 1 to 10 |
Pore Size | Monolith |
Temperature | 80°C |
Product Code | Brand | Particle Size | Diameter (Metric) | Length (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Particle Size | Diameter (Metric) | Length (Metric) | Price | Quantity & Availability | |||
11322023
|
Thermo Scientific™
164558 |
Monolith | 0.2 mm | 5 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
11312023
|
Thermo Scientific™
164557 |
Monolith | 0.2 mm | 50 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
11302023
|
Thermo Scientific™
164543 |
Monolith | 0.1 mm | 250 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
Description
For fast, high-resolution LC/MS separations in bottom-up protein identification, biomarker discovery, and systems biology applications, use Thermo Scientific™ PepSwift™ Monolithic Capillary LC Columns. Based on a polystyrene divinylbenzene copolymer, the monolithic structure offers a high-quality alternative to traditional microparticulate sorbents, providing important advantages to the chromatographic separation. High-sensitivity proteomics and biotech applications are easily performed using these columns.
Benefit from:- High-speed peptide and protein separations (<15 min)
- High sensitivity for LC/MS
- High column-to-column reproducibility
- Wide range of column I.D. and lengths available
- Superior lifetime
- Easy column installation using Thermo Scientific™ Dionex™ nanoViper™ Fingertight Fittings
- Precolumns available to preconcentrate and desalt samples
Specifications
4350 psi (300 bar) | |
1 to 10 | |
80°C | |
nanoLC | |
Reversed Phase |
Monolith | |
Monolith | |
PSDVB (Polystyrene Divinylbenzene) | |
Monolith Polymer | |
Reversed Phase |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.