missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PEPT2/SLC15A2 Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP1-47206
This item is not returnable.
View return policy
Description
PEPT2/SLC15A2 Polyclonal specifically detects PEPT2/SLC15A2 in Human, Porcine samples. It is validated for Peptide ELISA.
Specifications
| PEPT2/SLC15A2 | |
| Polyclonal | |
| Peptide ELISA Detection limit 1:8000 | |
| NP_066568.2 | |
| SLC15A2 | |
| Peptide with sequence C-QGNMIKLETKKTK corresponding to C-Terminus according to NP_066568.2. | |
| 0.1 mg | |
| Primary | |
| This antibody is specific to the C Terminus. | |
| Store at -20C. Avoid freeze-thaw cycles. |
| ELISA | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| Kidney H(+)/peptide cotransporter, kidney isoform, PEPT2FLJ33407, Peptide transporter 2, solute carrier family 15 (H+/peptide transporter), member 2, solute carrier family 15 member 2 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 6565 | |
| Human, Pig | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction