missing translation for 'onlineSavingsMsg'
Learn More

Perfluoro(1,3-dimethylcyclohexane), tech. 80%, remainder other fluorocarbons, Thermo Scientific™

Product Code. 11367575
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
Product Code. Quantity unitSize
11367575 25 g 25g
11377575 100 g 100g
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 11367575

Brand: Thermo Scientific Alfa Aesar L05726.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 335-27-3
Molecular Formula C8F16
Molecular Weight (g/mol) 400.062
MDL Number MFCD00001469
InChI Key LOQGSOTUHASIHI-UHFFFAOYSA-N
Synonym perfluoro-1,3-dimethylcyclohexane, perfluoro 1,3-dimethylcyclohexane, 1,1,2,2,3,3,4,5,5,6-decafluoro-4,6-bis trifluoromethyl cyclohexane, decafluoro-1,3-bis trifluoromethyl cyclohexane, cyclohexane, perfluoro-1,3-dimethyl, cyclohexane, 1,1,2,2,3,3,4,5,5,6-decafluoro-4,6-bis trifluoromethyl, cyclohexane, decafluoro-1,3-bis trifluoromethyl, hexadecafluoro 1,3-dimethylcyclohexane, flutec pp3, acmc-1cjoy
PubChem CID 78975
IUPAC Name 1,1,2,2,3,3,4,5,5,6-decafluoro-4,6-bis(trifluoromethyl)cyclohexane
SMILES C1(C(C(C(C(C1(F)F)(F)F)(F)F)(C(F)(F)F)F)(F)F)(C(F)(F)F)F

Specifications

Melting Point -70°C
Density 1.828
Boiling Point 101°C to 102°C
Refractive Index 1.2908
Quantity 25 g
Beilstein 2064147
Formula Weight 400.06
Percent Purity 80%
Grade Technical
Assay remainder other fluorocarbons
Chemical Name or Material Perfluoro(1,3-dimethylcyclohexane)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.