missing translation for 'onlineSavingsMsg'
Learn More

Perfluoro(tetradecahydrophenanthrene), mixture of isomers, Thermo Scientific Chemicals

Product Code. 11423148
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11423148 100 g 100g
11413148 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11423148 Supplier Thermo Scientific Alfa Aesar Supplier No. A18898.22
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Perfluoro(tetradecahydrophenanthrene) is used as a solvent in the preparation of co polymers obtained from emulsion polymerization of 2,3,3,3-tetrafluoroethylene and tetrafluoroethylene (TFE).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Perfluoro(tetradecahydrophenanthrene) is used as a solvent in the preparation of co polymers obtained from emulsion polymerization of 2,3,3,3-tetrafluoroethylene and tetrafluoroethylene (TFE).

Solubility
Immiscible with water.

Notes
Incompatible with lithium, sodium/sodium oxides, potassium and calcium.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 306-91-2
Molecular Formula C14F24
Molecular Weight (g/mol) 624.116
MDL Number MFCD00042596
InChI Key QKENRHXGDUPTEM-UHFFFAOYSA-N
Synonym perfluorophenanthrene, perfluoroperhydrophenanthrene, flutec pp11, perfluoro perhydrophenanthrene, tetracosafluorotetradecahydrophenanthrene, phenanthrene, tetracosafluorotetradecahydro, phenanthrene, 1,1,2,2,3,3,4,4,4a,4b,5,5,6,6,7,7,8,8,8a,9,9,10,10,10a-tetracosafluorotetradecahydro, flutec tm pp11 r, acmc-1clkp, dsstox_cid_27029
PubChem CID 78972
ChEBI CHEBI:39423
IUPAC Name 1,1,2,2,3,3,4,4,4a,4b,5,5,6,6,7,7,8,8,8a,9,9,10,10,10a-tetracosafluorophenanthrene
SMILES C12(C3(C(C(C(C1(C(C(C(C2(F)F)(F)F)(F)F)(F)F)F)(F)F)(F)F)(C(C(C(C3(F)F)(F)F)(F)F)(F)F)F)F)F

Specifications

Melting Point -20°C
Density 2.03
Boiling Point 215°C
Refractive Index 1.3348
Quantity 100 g
Solubility Information Immiscible with water.
Formula Weight 624.12
Chemical Name or Material Perfluoro(tetradecahydrophenanthrene), mixture of isomers

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.