missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Perforin Antibody (rPRF1/8058), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-23638-20ug
This item is not returnable.
View return policy
Description
Perforin Monoclonal antibody specifically detects Perforin in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)
Specifications
| Perforin | |
| Monoclonal | |
| 0.2 mg/mL | |
| Western Blot : 1-2 μg/mL, Immunohistochemistry-Paraffin : 1-2 μg/mL | |
| Cytolysin, FLH2, HPLH2, HPLH2lymphocyte pore forming protein, Lymphocyte pore-forming protein, MGC65093, P1PFN1, perforin 1 (pore forming protein), perforin-1, PFPcytolysin | |
| Recombinant human Perforin protein fragment (around aa 355-555) (exact sequence is proprietary) | |
| 20 μg | |
| Adaptive Immunity, Apoptosis, Immunology, Innate Immunity | |
| 5551 | |
| Store at 4°C. | |
| IgG1 κ |
| Western Blot, Immunohistochemistry (Paraffin) | |
| rPRF1/8058 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction