missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Periaxin Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93522-0.02ml
This item is not returnable.
View return policy
Description
Periaxin Polyclonal antibody specifically detects Periaxin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| Periaxin | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50-1:200 | |
| CMT4F, PRX | |
| A synthetic peptide corresponding to a sequence within amino acids 1250-1350 of human PRX (NP_870998.2). LGARARVGGEGAEEQPPGAERTFCLSLPDVELSPSGGNHAEYQVAEGEGEAGHKLKVRLPRFGLVRAKEGAEEGEKAKSPKLRLPRVGFSQSEMVTGEGSP | |
| 0.02 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 57716 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction