missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
The antibody is specific to PERK.
Phosphorylates the alpha subunit of eukaryotic translation-initiation factor 2 (EIF2), leading to its inactivation and thus to a rapid reduction of translational initiation and repression of global protein synthesis. Serves as a critical effector of unfolded protein response (UPR)-induced G1 growth arrest due to the loss of cyclin-D1 (CCND1). Involved in control of mitochondrial morphology and function.
Specifications
Specifications
| Antigen | PERK/EIF2AK3 |
| Applications | Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.46 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | EIF2AK3 |
| Gene Accession No. | Q9NZJ5, Q9Z2B5 |
| Gene Alias | EIF2AK3, HRI, HsPEK, Pancreatic eIF2 alpha kinase, PEK, PERK, WRS |
| Gene Symbols | EIF2AK3 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?